2,4-Dimethoxy-4'-hydroxybenzophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (2,4-Dimethoxyphenyl)(4-hydroxyphenyl)methanone

M-Clarity™ Qualitätsstufe = MQ100

542113-5G
SIAL542113-5GEA 44.8 EUR
SIAL542113-5G
2,4-Dimethoxy-4'-hydroxybenzophenon, Sigma-Aldrich®
2,4-Dimethoxy-4'-hydroxybenzophenon
Formel: C₁₅H₁₄O₄
Molecular Weight: 258,27 g/mol
MDL: MFCD00137850
CAS-Nummer: 41351-30-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR