3'-Brom-5'-chlor-2'-hydroxyacetophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-(3-Bromo-5-chloro-2-hydroxyphenyl)ethanone, 5-Chloro-3-bromo-2-hydroxyacetophenone

M-Clarity™ Qualitätsstufe = MQ100

542121-25G
SIAL542121-25GEA 309 EUR
SIAL542121-25G
3'-Brom-5'-chlor-2'-hydroxyacetophenon, Sigma-Aldrich®
3'-Brom-5'-chlor-2'-hydroxyacetophenon
Formel: C₈H₆BrClO₂
Molecular Weight: 249,49 g/mol
MDL: MFCD00857068
CAS-Nummer: 59443-15-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR