2-Chloro-3′,4′-dimethoxybenzil ≥97%, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-(2-Chlorophenyl)-2-(3,4-dimethoxyphenyl)ethane-1,2-dione

M-Clarity™ Qualitätsstufe = MQ100

542229-25G
SIAL542229-25GEA 52.5 EUR
SIAL542229-25G
2-Chloro-3′,4′-dimethoxybenzil ≥97%, Sigma-Aldrich®
2-Chloro-3′,4′-dimethoxybenzil

2-Chloro-3′,4′-dimethoxybenzil is a specific inhibitor of human carboxylesterase-2 (hCE-2).


  • Synonym: 1-(2-Chlorophenyl)-2-(3,4-dimethoxyphenyl)ethane-1,2-dione
  • Gene Information: human ACHE, BCHE and CES1


2-Chloro-3′,4′-dimethoxybenzil (2-Chloro-3,4-dimethoxybenzil) used in the synthesis of 2-(2-chlorophenyl)-3-(3,4-dimethoxyphenyl)quinoxaline and 3-(2-chlorophenyl)-4-(3,4-dimethoxyphenyl)-2,5-diphenylcyclopenta-2,4-dienone.


Verpackung: Packed 25 g in glass bottle.

Molecular Weight: 304,73 g/mol
Schmelzpunkt: 119…123 °C (Lit.)
CAS-Nummer: 56159-70-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR