3',5'-Dibrom-4'-hydroxyacetophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-(3,5-Dibromo-4-hydroxyphenyl)ethanone

M-Clarity™ Qualitätsstufe = MQ100

542245-5G
SIAL542245-5GEA 76 EUR
SIAL542245-5G
3',5'-Dibrom-4'-hydroxyacetophenon, Sigma-Aldrich®
3',5'-Dibrom-4'-hydroxyacetophenon
Formel: C₈H₆Br₂O₂
Molecular Weight: 293,9400 g/mol
MDL: MFCD00075779
CAS-Nummer: 2887-72-1
EINECS: 220-750-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR