Methyl-2,6-difluorbenzoat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl 2,6-difluorobenzoate

M-Clarity™ Qualitätsstufe = MQ100

542636-25G
SIAL542636-25GEA 70.1 EUR
SIAL542636-25G
Methyl-2,6-difluorbenzoat, Sigma-Aldrich®
Methyl-2,6-difluorbenzoat
Formel: C₈H₆F₂O₂
Molecular Weight: 172,13 g/mol
MDL: MFCD00051778
CAS-Nummer: 13671-00-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR