2-Fluorbenzolsulfonamid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Fluorbenzolsulfonsäureamid

M-Clarity™ Qualitätsstufe = MQ100

542717-5G
SIAL542717-5GEA 107 EUR
SIAL542717-5G
2-Fluorbenzolsulfonamid, Sigma-Aldrich®
2-Fluorbenzolsulfonamid
Formel: C₆H₆FNO₂S
Molecular Weight: 175,18 g/mol
MDL: MFCD00042283
CAS-Nummer: 30058-40-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR