3-Fluorbenzolsulfonamid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Fluorbenzolsulfonsäureamid

M-Clarity™ Qualitätsstufe = MQ100

542725-1G
SIAL542725-1GEA 42.1 EUR
SIAL542725-1G
3-Fluorbenzolsulfonamid, Sigma-Aldrich®
3-Fluorbenzolsulfonamid
Formel: C₆H₆FNO₂S
Molecular Weight: 175,18 g/mol
MDL: MFCD00042284
CAS-Nummer: 1524-40-9
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR