4-(Trifluormethyl)benzamidoxim, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

542822-1G
SIAL542822-1GEA 32.3 EUR
SIAL542822-1G
4-(Trifluormethyl)benzamidoxim, Sigma-Aldrich®
4-(Trifluormethyl)benzamidoxim
Formel: C₈H₇F₃N₂O
Molecular Weight: 204,15 g/mol
MDL: MFCD00053042
CAS-Nummer: 22179-86-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR