2-(Trifluormethoxy)phenylacetonitril, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-(Trifluormethoxy)benzylcyanid

M-Clarity™ Qualitätsstufe = MQ100

542881-1G
SIAL542881-1GEA 53.8 EUR
SIAL542881-1G
2-(Trifluormethoxy)phenylacetonitril, Sigma-Aldrich®
2-(Trifluormethoxy)phenylacetonitril
Formel: C₉H₆F₃NO
Molecular Weight: 201,15 g/mol
MDL: MFCD00236326
CAS-Nummer: 137218-25-8
UN: 3276
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR