1-Chlor-2-fluor-5-iodbenzol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Chlor-4-fluoriodbenzol, 2-Chlor-1-fluor-4-iodbenzol

M-Clarity™ Qualitätsstufe = MQ100

542903-25G
SIAL542903-25GEA 54.3 EUR
SIAL542903-25G
1-Chlor-2-fluor-5-iodbenzol, Sigma-Aldrich®
1-Chlor-2-fluor-5-iodbenzol
Formel: C₆H₃ClFI
Molecular Weight: 256,45 g/mol
MDL: MFCD00042210
CAS-Nummer: 156150-67-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR