N-Benzyloxycarbonyl-L-alaninol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Z-Ala-ol, (S)-Benzyl (1-hydroxypropan-2-yl)carbamate, (S)-(2-Hydroxy-1-methylethyl)carbamic acid benzyl ester, N-Cbz-L-Alaninol

M-Clarity™ Qualitätsstufe = MQ100

543217-5G
SIAL543217-5GEA 136 EUR
SIAL543217-5G
N-Benzyloxycarbonyl-L-alaninol, Sigma-Aldrich®
N-Cbz-L-Alaninol
Formel: C₁₁H₁₅NO₃
Molecular Weight: 209,25 g/mol
MDL: MFCD00672530
CAS-Nummer: 66674-16-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR