4-(4-Brombenzyloxy)benzaldehyd, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-((4-Bromobenzyl)oxy)benzaldehyde

M-Clarity™ Qualitätsstufe = MQ100

543330-1G
SIAL543330-1GEA 53.7 EUR
SIAL543330-1G
4-(4-Brombenzyloxy)benzaldehyd, Sigma-Aldrich®
4-(4-Brombenzyloxy)benzaldehyd
Formel: C₁₄H₁₁BrO₂
Molecular Weight: 291,1399 g/mol
MDL: MFCD02091001
CAS-Nummer: 149833-95-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR