(2-Hydroxyethyl)hydrazin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: β-Hydroxyethylhydrazine

54340-100ML
SIAL54340-100MLEA 127 EUR
SIAL54340-100ML
(2-Hydroxyethyl)hydrazin, Sigma-Aldrich®
(2-Hydroxyethyl)hydrazine
Formel: C₂H₈N₂O
MDL: MFCD00007623
CAS-Nummer: 109-84-2
EINECS: 203-711-6
UN: 2810
ADR: 6.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR