6-Fluor-3-formylchromon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 6-Fluorochromone-3-carboxaldehyde, 6-Fluoro-4-oxo-4H-chromene-3-carbaldehyde

M-Clarity™ Qualitätsstufe = MQ100

543624-1G
SIAL543624-1GEA 37.4 EUR
SIAL543624-1G
6-Fluor-3-formylchromon, Sigma-Aldrich®
6-Fluor-3-formylchromon
Formel: C₁₀H₅FO₃
Molecular Weight: 192,15 g/mol
MDL: MFCD00139060
CAS-Nummer: 69155-76-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR