5-(5-Fluor-2-hydroxyphenyl)isoxazol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Fluoro-2-(5-isoxazolyl)phenol, 5-(5-Fluoro-2-hydroxyphenyl)isoxazole

M-Clarity™ Qualitätsstufe = MQ100

543683-1G
SIAL543683-1GEA 30.2 EUR
SIAL543683-1G
5-(5-Fluor-2-hydroxyphenyl)isoxazol, Sigma-Aldrich®
5-(5-Fluor-2-hydroxyphenyl)isoxazol
Formel: C₉H₆FNO₂
MDL: MFCD03070563
CAS-Nummer: 288401-62-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR