6-Fluorchromon-2-carbonsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 6-Fluoro-4-oxo-4H-chromene-2-carboxylic acid

M-Clarity™ Qualitätsstufe = MQ100

543764-1G
SIAL543764-1GEA 52 EUR
SIAL543764-1G
6-Fluorchromon-2-carbonsäure, Sigma-Aldrich®
6-Fluorchromon-2-carbonsäure
Formel: C₁₀H₅FO₄
Molecular Weight: 208,15 g/mol
MDL: MFCD03070543
CAS-Nummer: 99199-59-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR