5'-Fluor-2'-hydroxy-3'-nitroacetophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-(5-Fluoro-2-hydroxy-3-nitrophenyl)ethanone

M-Clarity™ Qualitätsstufe = MQ100

543780-1G
SIAL543780-1GEA 48.6 EUR
SIAL543780-1G
5'-Fluor-2'-hydroxy-3'-nitroacetophenon, Sigma-Aldrich®
5'-Fluor-2'-hydroxy-3'-nitroacetophenon
Formel: C₈H₆FNO₄
Molecular Weight: 199,136 g/mol
MDL: MFCD03094007
CAS-Nummer: 70978-39-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR