4-Hydroxycarbazol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Carbazolol, Carbazol-2-ol

MQ100

543896-5G
SIAL543896-5GEA 66.5 EUR
SIAL543896-5G
4-Hydroxycarbazol, Sigma-Aldrich®
4-Hydroxycarbazole
Formel: C₁₂H₉NO
Molecular Weight: 183.21 g/mol
MDL: MFCD02178385
CAS-Nummer: 52602-39-8
EINECS: 258-034-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR