2-Fluor-5-nitrobenzonitril, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

544221-1G
SIAL544221-1GEA 39.3 EUR
SIAL544221-1G
2-Fluor-5-nitrobenzonitril, Sigma-Aldrich®
2-Fluor-5-nitrobenzonitril
Formel: C₇H₃FN₂O₂
Molecular Weight: 166,11 g/mol
Schmelzpunkt: 79 °C
MDL: MFCD00042299
CAS-Nummer: 17417-09-3
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR