1,5-Dimethoxypentan, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Pentamethylene glycol dimethyl ether, 1,5-Pentanediol dimethyl ether

M-Clarity™ Qualitätsstufe = MQ100

544469-25G
SIAL544469-25GEA 297 EUR
SIAL544469-25G
1,5-Dimethoxypentan, Sigma-Aldrich®
1,5-Dimethoxypentane
Formel: C₇H₁₆O₂
Molecular Weight: 132,200 g/mol
MDL: MFCD03613608
CAS-Nummer: 111-89-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR