5-Brom-2-methoxybenzolsulfonylchlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Brom-2-methoxybenzolsulfonsäurechlorid, 5-Brom-2-methoxybenzolsulfochlorid

544515-5G
SIAL544515-5GEA 137 EUR
SIAL544515-5G
5-Brom-2-methoxybenzolsulfonylchlorid, Sigma-Aldrich®
5-Brom-2-methoxybenzolsulfonylchlorid
Formel: C₇H₆BrClO₃S
Molecular Weight: 285,55 g/mol
Schmelzpunkt: 107…110 °C
MDL: MFCD00051768
CAS-Nummer: 23095-05-8
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR