4-Methoxybenzolsulfonamid, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
544655-5G
SIAL544655-5GEA
74.9
EUR
SIAL544655-5G
4-Methoxybenzolsulfonamid, Sigma-Aldrich®
4-Methoxybenzolsulfonamid
Formel:
C₇H₉NO₃S Molecular Weight: 187,22 g/mol |
MDL:
MFCD00025392 CAS-Nummer: 1129-26-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...