Di-tert-butylsilan, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

545236-5ML
SIAL545236-5MLEA 125 EUR
SIAL545236-5ML
Di-tert-butylsilan, Sigma-Aldrich®
Di-tert-butylsilan
Formel: C₈H₂₀Si
Molecular Weight: 144,33 g/mol
MDL: MFCD00270996
CAS-Nummer: 30736-07-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR