3-Octin-1-ol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

545430-5G
SIAL545430-5GEA 73.1 EUR
SIAL545430-5G
3-Octin-1-ol, Sigma-Aldrich®
3-Octin-1-ol
Formel: C₈H₁₄O
MDL: MFCD00039552
CAS-Nummer: 14916-80-4
EINECS: 238-986-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR