5-Chlor-2-methylbenzothiazol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

545775-5G
SIAL545775-5GEA 51 EUR
SIAL545775-5G
5-Chlor-2-methylbenzothiazol, Sigma-Aldrich®
5-Chloro-2-methylbenzothiazole
Formel: C₈H₆ClNS
Molecular Weight: 183,66 g/mol
MDL: MFCD00022881
CAS-Nummer: 1006-99-1
EINECS: 213-746-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR