4-Chlor-2-hydroxy-5-sulfamoylbenzoesäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

545805-25G
SIAL545805-25GEA 0 EUR
SIAL545805-25G
4-Chlor-2-hydroxy-5-sulfamoylbenzoesäure, Sigma-Aldrich®
4-Chlor-2-hydroxy-5-sulfamoylbenzoesäure
Formel: C₇H₆ClNO₅S
Molecular Weight: 251,65 g/mol
CAS-Nummer: 14556-98-0
EINECS: 238-602-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR