2-Methyl-5-nitrobenzothiazol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

545821-5G
SIAL545821-5GEA 240 EUR
SIAL545821-5G
2-Methyl-5-nitrobenzothiazol, Sigma-Aldrich®
2-Methyl-5-nitrobenzothiazole
Formel: C₈H₆N₂O₂S
MDL: MFCD00602735
CAS-Nummer: 2941-66-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR