(4-Fluorphenoxy)essigsäurehydrazid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (4-Fluorphenoxy)acethydrazid, (4-Fluorphenoxy)acetohydrazid

M-Clarity™ Qualitätsstufe = MQ100

545937-5G
SIAL545937-5GEA 30.8 EUR
SIAL545937-5G
(4-Fluorphenoxy)essigsäurehydrazid, Sigma-Aldrich®
(4-Fluorphenoxy)essigsäurehydrazid
Formel: (C₆H₄F)OCH₂CONHNH₂
Molecular Weight: 184,17 g/mol
MDL: MFCD00082651
CAS-Nummer: 1737-62-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR