6-(Trifluormethyl)uracil, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-(Trifluormethyl)uracil, 2,4-Dihydroxy-6-(trifluormethyl)pyrimidin, 2,6-Dihydroxy-4-(trifluormethyl)pyrimidin

M-Clarity™ Qualitätsstufe = MQ100

546011-5G
SIAL546011-5GEA 161 EUR
SIAL546011-5G
6-(Trifluormethyl)uracil, Sigma-Aldrich®
6-(Trifluormethyl)uracil
Formel: C₅H₃F₃N₂O₂
Molecular Weight: 180,09 g/mol
MDL: MFCD01011762
CAS-Nummer: 672-45-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR