3-(2-Benzothiazolyl)-7-(diethylamino)cumarin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-(Benzo[d]thiazol-2-yl)-7-(diethylamino)-2H-chromen-2-one, Coumarin 6

M-Clarity™ Qualitätsstufe = MQ100

546283-100MG
SIAL546283-100MGEA 138 EUR
SIAL546283-100MG
3-(2-Benzothiazolyl)-7-(diethylamino)cumarin, Sigma-Aldrich®
3-(2-Benzothiazolyl)-7-(diethylamino)cumarin
Formel: C₂₀H₁₈N₂O₂S
Molecular Weight: 350,4341 g/mol
MDL: MFCD00041869
CAS-Nummer: 38215-36-0
EINECS: 253-830-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR