3-(4-Brombenzoyl)propionsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-(4-Bromophenyl)-4-oxobutanoic acid

M-Clarity™ Qualitätsstufe = MQ100

546291-25G
SIAL546291-25GEA 173 EUR
SIAL546291-25G
3-(4-Brombenzoyl)propionsäure, Sigma-Aldrich®
3-(4-Brombenzoyl)propionsäure
Formel: C₁₀H₉BrO₃
Molecular Weight: 257,08 g/mol
MDL: MFCD00016563
CAS-Nummer: 6340-79-0
EINECS: 228-732-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR