5-Chlor-2-nitrobenzamid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Chlor-2-nitrobenzoesäureamid, 3-Chlor-6-nitrobenzoesäureamid

M-Clarity™ Qualitätsstufe = MQ100

546542-10G
SIAL546542-10GEA 133 EUR
SIAL546542-10G
5-Chlor-2-nitrobenzamid, Sigma-Aldrich®
5-Chlor-2-nitrobenzamid
Formel: C₇H₅ClN₂O₃
Molecular Weight: 200,58 g/mol
MDL: MFCD00017509
CAS-Nummer: 40763-96-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR