3,5-Difluor-DL-phenylalanin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

546550-500MG
SIAL546550-500MGEA 56.4 EUR
SIAL546550-500MG
3,5-Difluor-DL-phenylalanin, Sigma-Aldrich®
3,5-Difluor-DL-phenylalanin
Formel: C₉H₉F₂NO₂
Molecular Weight: 201,1701 g/mol
MDL: MFCD00061308
CAS-Nummer: 32133-37-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR