3-Brom-α,α,α,4-tetrafluortoluol, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 3-Brom-4-fluorbenzotrifluorid, 1-Brom-2-fluor-5-(trifluormethyl)benzol

M-Clarity™ Qualitätsstufe = MQ100

546577-5G
SIAL546577-5GEA 88.5 EUR
SIAL546577-5G
3-Brom-α,α,α,4-tetrafluortoluol, Sigma-Aldrich®
3-Brom-α,α,α,4-tetrafluortoluol
Formel: C₇H₃BrF₄
Molecular Weight: 243 g/mol
MDL: MFCD00040938
CAS-Nummer: 68322-84-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR