trans-2-Octen-1-ol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: trans-1-Hydroxy-2-octen, trans-Oct-2-en-1-ol

M-Clarity™ Qualitätsstufe = MQ100

547115-25G
SIAL547115-25GEA 120 EUR
SIAL547115-25G
trans-2-Octen-1-ol, Sigma-Aldrich®
trans-2-Octen-1-ol
Formel: C₈H₁₆O
Molecular Weight: 128,21 g/mol
MDL: MFCD00014057
CAS-Nummer: 18409-17-1
EINECS: 242-288-2
UN: 1987
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR