Tris[N,N-bis(trimethylsilyl)amid]lanthan(III), Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

547778-1G
SIAL547778-1GEA 137 EUR
SIAL547778-1G
Tris[N,N-bis(trimethylsilyl)amid]lanthan(III), Sigma-Aldrich®
Tris[N,N-bis(trimethylsilyl)amid]lanthan(III)
Formel: C₁₈H₅₄LaN₃Si₆
Molecular Weight: 620,06 g/mol
MDL: MFCD00271009
CAS-Nummer: 175923-07-6
UN: 3396
ADR: 4.3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR