Tris[N,N-bis(trimethylsilyl)amid]europium(III), Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

547816-1G
SIAL547816-1GEA 0 EUR
SIAL547816-1G
Tris[N,N-bis(trimethylsilyl)amid]europium(III), Sigma-Aldrich®
Tris[N,N-bis(trimethylsilyl)amid]europium(III)
Formel: C₁₈H₅₄EuN₃Si₆
MDL: MFCD03411248
CAS-Nummer: 35789-02-7
UN: 3396
ADR: 4.3 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR