Tris[N,N-bis(trimethylsilyl)amid]gadolinium(III), Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

547824-1G
SIAL547824-1GEA 161 EUR
SIAL547824-1G
Tris[N,N-bis(trimethylsilyl)amid]gadolinium(III), Sigma-Aldrich®
Tris[N,N-bis(trimethylsilyl)amid]gadolinium(III)
Formel: C₁₈H₅₄GdN₃Si₆
Molecular Weight: 638,41 g/mol
MDL: MFCD03427091
CAS-Nummer: 35789-03-8
UN: 3396
ADR: 4.3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR