Tris[N,N-bis(trimethylsilyl)amid]samarium(III), Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

547980-1G
SIAL547980-1GEA 200 EUR
SIAL547980-1G
Tris[N,N-bis(trimethylsilyl)amid]samarium(III), Sigma-Aldrich®
Tris[N,N-bis(trimethylsilyl)amid]samarium(III)
Formel: C₁₈H₅₄N₃Si₆Sm
MDL: MFCD03427097
CAS-Nummer: 35789-01-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR