3-(Trifluormethyl)benzamid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-(Trifluormethyl)benzoesäureamid

M-Clarity™ Qualitätsstufe = MQ100

548146-5G
SIAL548146-5GEA 30.1 EUR
SIAL548146-5G
3-(Trifluormethyl)benzamid, Sigma-Aldrich®
3-(Trifluormethyl)benzamid
Formel: C₈H₆F₃NO
Molecular Weight: 189,14 g/mol
MDL: MFCD00014802
CAS-Nummer: 1801-10-1
EINECS: 217-289-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR