6-Amino-3-brom-2-methylpyridin, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 5-Brom-6-methylpyridin-2-amin, 2-Amino-5-brom-6-methylpyridin, 3-Brom-2-methyl-6-pyridylamin, 3-Brom-2-methylpyridin-6-amin, 5-Brom-6-methyl-2-pyridylamin, 6-Amino-3-brom-2-picolin, 2-Amino-5-brom-6-picolin

M-Clarity™ Qualitätsstufe = MQ100

548405-5G
SIAL548405-5GEA 42.2 EUR
SIAL548405-5G
6-Amino-3-brom-2-methylpyridin, Sigma-Aldrich®
6-Amino-3-brom-2-methylpyridin
Formel: C₆H₇BrN₂
Molecular Weight: 187,04 g/mol
MDL: MFCD00068230
CAS-Nummer: 42753-71-9
EINECS: 255-927-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR