3-Methylbenzamidoxim, Sigma-Aldrich®

Lieferant: Merck

Synonyme: N-Hydroxy-3-methylbenzimidamide

M-Clarity™ Qualitätsstufe = MQ100

548596-1G
SIAL548596-1GEA 47.9 EUR
SIAL548596-1G
3-Methylbenzamidoxim, Sigma-Aldrich®
3-Methylbenzamidoxim
Formel: C₈H₁₀N₂O
Molecular Weight: 150,18 g/mol
MDL: MFCD00833266
CAS-Nummer: 40067-82-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR