3-(4-Methylphenoxy)propionsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-(p-Tolyloxy)propanoic acid

M-Clarity™ Qualitätsstufe = MQ100

548863-25G
SIAL548863-25GEA 0 EUR
SIAL548863-25G
3-(4-Methylphenoxy)propionsäure, Sigma-Aldrich®
3-(4-Methylphenoxy)propionsäure
Formel: C₁₀H₁₂O₃
Molecular Weight: 180,2 g/mol
MDL: MFCD00220218
CAS-Nummer: 25173-37-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR