2,5-Bis(trifluormethyl)benzolsulfonylchlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,5-Bis(trifluormethyl)benzolsulfonsäurechlorid, 2,5-Bis(trifluormethyl)benzol-1-sulfonylchlorid, 2,5-Bis(trifluormethyl)benzolsulfochlorid

M-Clarity™ Qualitätsstufe = MQ100

549002-1G
SIAL549002-1GEA 53.2 EUR
SIAL549002-1G
2,5-Bis(trifluormethyl)benzolsulfonylchlorid, Sigma-Aldrich®
2,5-Bis(trifluormethyl)benzolsulfonylchlorid
Formel: C₈H₃ClF₆O₂S
Molecular Weight: 312,62 g/mol
MDL: MFCD03094042
CAS-Nummer: 351003-22-0
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR