3-Brom-α,α,α,2-tetrafluortoluol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Brom-2-fluor-3-(trifluormethyl)benzol, 3-Brom-2-fluorbenzotrifluorid

M-Clarity™ Qualitätsstufe = MQ100

549010-5G
SIAL549010-5GEA 107 EUR
SIAL549010-5G
3-Brom-α,α,α,2-tetrafluortoluol, Sigma-Aldrich®
3-Brom-α,α,α,2-tetrafluortoluol
Formel: C₇H₃BrF₄
Molecular Weight: 243 g/mol
MDL: MFCD00070812
CAS-Nummer: 144584-67-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR