5-Brom-α,α,α,2-tetrafluortoluol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Brom-2-fluorbenzotrifluorid, 1-Brom-4-fluor-3-(trifluormethyl)benzol

M-Clarity™ Qualitätsstufe = MQ200

549096-5G
SIAL549096-5GEA 68.7 EUR
SIAL549096-5G
5-Brom-α,α,α,2-tetrafluortoluol, Sigma-Aldrich®
5-Brom-α,α,α,2-tetrafluortoluol
Formel: C₇H₃BrF₄
Molecular Weight: 243 g/mol
MDL: MFCD00042493
CAS-Nummer: 393-37-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR