3-Cyan-4-methylanilin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Amino-2-methylbenzonitrile

M-Clarity™ Qualitätsstufe = MQ100

549584-25G
SIAL549584-25GEA 111 EUR
SIAL549584-25G
3-Cyan-4-methylanilin, Sigma-Aldrich®
3-Cyan-4-methylanilin
Formel: C₈H₈N₂
Molecular Weight: 132,16 g/mol
MDL: MFCD00017608
CAS-Nummer: 50670-64-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR