4-(Methylthio)phenol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

550426-25G
SIAL550426-25GEA 80.9 EUR
SIAL550426-25G
4-(Methylthio)phenol, Sigma-Aldrich®
4-(Methylthio)phenol
Formel: C₇H₈OS
MDL: MFCD00002351
CAS-Nummer: 1073-72-9
EINECS: 214-031-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR