4-Brom-4'-fluorbiphenyl, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Brom-4-(4-fluorphenyl)benzol

M-Clarity™ Qualitätsstufe = MQ100

550450-500MG
SIAL550450-500MGEA 30.5 EUR
SIAL550450-500MG
4-Brom-4'-fluorbiphenyl, Sigma-Aldrich®
4-Brom-4'-fluorbiphenyl
Formel: C₁₂H₈BrF
Molecular Weight: 251,1 g/mol
MDL: MFCD00017954
CAS-Nummer: 398-21-0
EINECS: 206-909-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR