3-Oxocyclohexancarbonsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

550515-1G
SIAL550515-1GEA 238 EUR
SIAL550515-1G
3-Oxocyclohexancarbonsäure, Sigma-Aldrich®
3-Oxocyclohexanecarboxylic acid
Formel: C₇H₁₀O₃
Molecular Weight: 142,1525 g/mol
MDL: MFCD01320172
CAS-Nummer: 16205-98-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR